Learn More
Spectrum Labs SPECTRUM CLEARBATH ALGAE INHIBITOR, 57.5ML
Product Code. 15193275
SPECTRUM CLEARBATH ALGAE INHIBITOR, 57.5ML
Product Code. 15193275 Supplier Spectrum Labs Supplier No. 105535
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
SPECTRUM CLEARBATH ALGAE INHIBITOR, 57.5ML
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Clear Bath
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H360FD-May damage fertility. May damage the unborn child.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P233-Keep container tightly closed.
  • P271-Use only outdoors or in a well-ventilated area.
  • P242-Use only non-sparking tools.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash thoroughly after handling.