Learn More
Spectrum™ Spectra/Por™ Weighted Paired Closures
Product Code. 10423404
Change view
Click to view available options
Diameter (Metric):
12 mm
23 mm
35 mm
55 mm
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10423404 12 mm Pack of 20
11540910 23 mm Pack of 20
11550910 35 mm Pack of 20
11560910 55 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10423404 Supplier Spectrum™ Supplier No. 132749
  • ONLINE EXCLUSIVE PRICE
    321.00€
    221.00€ / Pack of 20
    Save 100.00€
    31% Off
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
  • 10 pairs (20 closures)/pkg
Specifications
Product Type Dialysis Tubing Closure
Membrane Regenerated Cellulose
Material Polypropylene
Diameter (Metric) 12 mm
Autoclavable Autoclavable
Closure Type Paired
Color Orange or White
Temperature Range (English) 32°F to 250°F
Temperature Range (Metric) 0°C to 121°C
Quantity 20 Pack
For Use With (Application) Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal