missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermine synthase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15979-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).Specifications
| Spermine synthase | |
| Polyclonal | |
| Unconjugated | |
| SMS | |
| MRSR, SMS, Spermine synthase, SPMSY, SpS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20603, 363469, 6611 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52788, P97355 | |
| Sms | |
| Spermine synthase Fusion Protein Ag8767 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction