Learn More
Spermine synthase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15979-1-AP-20UL
 View more versions of this product
Product Code. 16875404
  • 145.00€ / 20µL
Description
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
Specifications
Spermine synthase
Polyclonal
Unconjugated
SMS
MRSR, SMS, Spermine synthase, SPMSY, SpS
Rabbit
Antigen Affinity Chromatography
RUO
20603, 363469, 6611
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52788, P97355
Sms
Spermine synthase Fusion Protein Ag8767