missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-22974CL1
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
| sphingosine kinase 1 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8877 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction