missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike RBD Antibody (2847A) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB11198JF585
This item is not returnable.
View return policy
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry,Western Blot
Specifications
| Spike RBD | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Western Blot | |
| 2847A | |
| 50mM Sodium Borate | |
| CHO-derived sars-cov-2 Spike RBD protein, Arg319-Phe541, Accession # YP_009724390.1 | |
| RUO | |
| SARS-CoV-2 | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction