Learn More
MP Biomedicals™ SPINeasy RNA Kit for Bacteria
Product Code. 17782438 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
50 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17782438 50 Preps 50 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17782438 Supplier MP Biomedicals™ Supplier No. 116541050
  • ONLINE EXCLUSIVE PRICE
    544.00€
    517.65€ / 50 sets
    Save 26.35€
    4% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SPINeasy RNA Kit for Bacteria - Quick and convenient purification of total RNA from gram-positive and gram-negative bacteria using spin column technology.
  • SPINeasy RNA Kit for Bacteria is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from gram-positive and gram-negative bacteria. Included in the kit is our specially formulated RNASS solution that stabilizes and protects RNA in bacteria samples.
  • The use of Lysis Buffer R and Lysing Matrix B in combination with FastPrep™ instruments from MP Biomedicals enables highly efficient lysis of bacterial samples within seconds.
  • With a simple workflow, the kit allows multiple samples to be processed:
    • Simultaneously
    • Without the use of toxic substances such as phenol and chloroform.
  • Total RNA of high quality and integrity can be purified typically:
    • Within 40 minutes
    • Is immediately available for RT-PCR and other downstream applications.
Specifications
Buffer Lysis Buffer
Isolation Technology Silica Spin Column
Purification Time 40 min.
Species E. coli
Sample Type Bacteria
Quantity 50 Preps
compliance-icons compliance-icons
Product Identifier
  • SPINeasy RNA Kit for Bacteria (With Lysing Matrix)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Manganese Sulphate Monohydrate, CalciumChloride, GuanidineHydrochloride