Learn More
Euro Protection™ Spinelle Safety Shoes
Product Code. 12084405
Change view
Click to view available options
Size:
41
43
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12084405 41 Each
12004415 43 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12084405 Supplier Euro Protection™ Supplier No. 9SPIL41
Euro Protection™ Spinelle Safety Shoes are breathable, padded, and feature a steel toe cap for protection.
  • Breathable upper.
  • Metal ventilation eyelets on the inner side.
  • Padded collar and tongue for snug fit.
  • Suede leather and mesh combination upper.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
  • Steel toe cap.
  • Stainless steel puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.
Specifications
Color Gray, Orange
Liner Material Breathable 3D mesh lining
Material Leather
Size 41
Sole Features Removable, textile and foam insole, stainless steel, puncture-resistant sole, injected polyurethan midsole, polyurethane outsole.
Sole Material Polyurethane, Stainless Steel
Toe Material Steel
Weight (Metric) 583 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Shoe