Learn More
Lenz Laborglasintrumente Spiral Condensers, with Glass Olives
Product Code. 10737331
Change view
Click to view available options
Length (Metric):
400 mm
500 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10737331 500 mm Each
10601902 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10737331 Supplier Lenz Laborglasintrumente Supplier No. 05233504
  • ONLINE EXCLUSIVE PRICE
    190.00€
    142.58€ / Pack of 1
    Save 47.42€
    24% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Spiral condensers for cooling condensates
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Length (Metric) 500 mm
Standard Taper Joint No NS 29/32