missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPOCK3 Monoclonal Antibody (330403)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561143
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Proteoglycans, which consist of a core protein and covalently linked glycosaminoglycans, are components of the extracellular matrix. SPOCK3 encodes a member of a novel Ca(2+)-binding proteoglycan family.
Specifications
| SPOCK3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8BKV0 | |
| Spock3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Testican 3/SPOCK3, Val26-Ile436. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA | |
| 330403 | |
| Unconjugated | |
| Spock3 | |
| 2900045C01Rik; AI428471; HSAJ1454; mKIAA4039; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 3; SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; SPOCK3; TES-3; testican 3; testican-3; TICN3; UNQ409/PRO771 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 72902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction