missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPON1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547457
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SPON1 is a cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the out growth of neurites in vitro. SPON1 may contribute to the growth and guidance of axons in both the spinal cord and the PNS. Is known to be a major factor for vascular smooth muscle cells.
Specifications
| SPON1 | |
| Polyclonal | |
| Unconjugated | |
| SPON1 | |
| AI666765; AW455831; BC020531; D330035F22Rik; FSP; f-spondin; KIAA0762; MGC10724; Spon1; spondin 1; spondin 1, (f-spondin) extracellular matrix protein; spondin 1, extracellular matrix protein; spondin-1; Sponf; Vascular smooth muscle cell growth-promoting factor; VSGP; VSGP/F-spondin | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10418 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9HCB6 | |
| SPON1 | |
| Mouse myeloma cell line NS0-derived recombinant human F-Spondin/SPON1 Phe29-Cys807. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction