missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRED1 Antibody (M23-P2G3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 351.00€
Specifications
| Antigen | SPRED1 |
|---|---|
| Clone | M23-P2G3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604578
|
Novus Biologicals
NBP2-50308 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604008
|
Novus Biologicals
NBP2-50308-0.025ml |
0.025 mL |
190.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPRED1 Monoclonal antibody specifically detects SPRED1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SPRED1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS | |
| 161742 | |
| IgG1 κ | |
| Protein A purified |
| M23-P2G3 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation | |
| Peptide Sequence YVPLRMCHR (amino acids 420 - 428) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title