missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRTN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5107278
Additional Details : Weight : 0.01000kg
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 92%; Rat: 100%
SPRTN is a regulator of UV-induced DNA damage response: acts as a 'reader' of ubiquitinated PCNA that enhances RAD18-mediated PCNA ubiquitination and translesion DNA synthesis. SPRTN is recruited to sites of UV damage and interacts with ubiquitinated PCNA and RAD18, the E3 ubiquitin ligase that monoubiquitinates PCNA. It facilitates chromatin association of RAD18 and is required for efficient PCNA monoubiquitination, promoting a feed-forward loop to enhance PCNA ubiquitination and translesion DNA synthesis. It is also required for localization of DNA polymerase eta (POLH) to sites of UV damage.Specifications
SPRTN | |
Polyclonal | |
Unconjugated | |
SPRTN | |
C1orf124; DDDL1880; dJ876B10.3; DNA damage protein targeting VCP; DNA damage-targeting VCP (p97) adaptor; DVC1; Gm505; PRO4323; protein with SprT-like domain at the N terminus; RGD1559496; spartan; sprT-like domain-containing protein Spartan; SprT-like N-terminal domain; Sprtn; UNQ1880/PRO4323; zinc finger RAD18 domain-containing protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83932 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q9H040 | |
SPRTN | |
Synthetic peptide located within the following region: ATNREPSAHD YWWAEHQKTC GGTYIKIKEP ENYSKKGKGK AKLGKEPVLA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |