missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRTN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546262
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QIKSSGNDPK YSTTTAQNSS SSSSQSKMVN CPVCQNEVLE SQINEHLDWC Sequence homology: Human: 100%
SPRTN Polyclonal antibody specifically detects SPRTN in Human samples. It is validated for Western Blot.Specifications
SPRTN | |
Polyclonal | |
Unconjugated | |
SPRTN | |
C1orf124; DDDL1880; dJ876B10.3; DNA damage protein targeting VCP; DNA damage-targeting VCP (p97) adaptor; DVC1; Gm505; PRO4323; protein with SprT-like domain at the N terminus; RGD1559496; spartan; sprT-like domain-containing protein Spartan; SprT-like N-terminal domain; Sprtn; UNQ1880/PRO4323; zinc finger RAD18 domain-containing protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83932 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H040 | |
SPRTN | |
Synthetic peptide directed towards the C-terminal of human SPRTN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |