Learn More
SPRYD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558882
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LGEEVRLHLN AELGREDDSV MMVDSYEDEW GRLHDVRVCG TLLEYLGKGK SIVDVGLAQA RHPLSTRSHY FEVEIVDPGE KCYIAL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The SPRY domain-containing protein 3 (SPRYD3) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. Little is known of the function of the SPRYD3 protein.Specifications
SPRYD3 | |
Polyclonal | |
Unconjugated | |
SPRYD3 | |
BC008150; RGD1561500; SPRY domain containing 3; SPRY domain-containing protein 3; SPRYD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84926 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCJ5 | |
SPRYD3 | |
Recombinant protein corresponding to Human SPRYD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only