Learn More
Invitrogen™ SR-BI Recombinant Rabbit Monoclonal Antibody (24GB1395)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557705
 View more versions of this product
Product Code. 30275413
  • 198.00€ / 20µL
Description
SR-BI Recombinant Monoclonal Antibody for Western Blot, ICC/IF
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SR-BI
Recombinant Monoclonal
Unconjugated
SCARB1
AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII
Rabbit
Affinity chromatography
RUO
25073, 949
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1395
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P97943, Q8WTV0
SCARB1
A synthesized peptide derived from human Scavenging Receptor SR-BI (aa 80-110).
20 μL
Primary
Human, Rat