missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRC3 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5148956
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
Spezifikation
| SRC3 | |
| Polyclonal | |
| Unconjugated | |
| NCOA3 | |
| 2010305B15Rik; Actr; AIB1; AIB-1; amplified in breast cancer 1 protein; amplified in breast cancer-1 protein homolog; AW321064; BHLHE42; CAGH16; CBP-interacting protein; class E basic helix-loop-helix protein 42; CTG26; KAT13B; MGC141848; NCOA3; NCoA-3; Nuclear receptor coactivator 3; OTTHUMP00000031716; p/Cip; pCip; Rac3; RAC-3; Receptor-associated coactivator 3; RP5-1049G16.3; Src3; SRC-3; steroid receptor coactivator protein 3; Thyroid hormone receptor activator molecule 1; TNRC14; TNRC16; Tram1; Tram-1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17979, 8202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O09000, Q9Y6Q9 | |
| NCOA3 | |
| E. coli-derived recombinant human NCOA3, Met673-Val856. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur