missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRCRB4D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12628-50ul
This item is not returnable.
View return policy
Description
SRCRB4D Polyclonal antibody specifically detects SRCRB4D in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SRCRB4D | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| S4D-SRCRB, S4-SRCRB, scavenger receptor cysteine rich domain containing, group B (4 domains), scavenger receptor cysteine-rich domain-containing group B protein, scavenger receptor cysteine-rich protein SRCRB-S4D, SRCRB-S4D, SSC4D | |
| Produced in rabbits immunized with E. coli-derived Human SRCRB4D fragment | |
| 50 μg | |
| Cell Biology | |
| 136853 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction