missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRSF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562086
This item is not returnable.
View return policy
Description
Immunogen sequence: LKVDNLTYRT SPDSLRRVFE KYGRVGDVYI PR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
The protein encoded by SC35 is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two transcript variants encoding the same protein and one non-coding transcript variant have been found for SC35. In addition, a pseudogene of SC35 has been found on chromosome 11.
Specifications
| SRSF2 | |
| Polyclonal | |
| Unconjugated | |
| SRSF2 | |
| D11Wsu175e; MRF-1; myelin regulatory factor 1; Pr264; Protein PR264; Putative myelin regulatory factor 1; SC35; SC-35; serine and arginine rich splicing factor 2; serine/arginine-rich splicing factor 2; Sfrs10; Sfrs2; SFRS2A; splicing component, 35 kDa; Splicing factor SC35; splicing factor, arginine/serine-rich 2; splicing factor, arginine/serine-rich 2 (SC-35); SR splicing factor 2; SRp30b; Srsf2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6427 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01130 | |
| SRSF2 | |
| Recombinant protein corresponding to Human SRSF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction