Learn More
SSBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12212-1-AP-20UL
 View more versions of this product
Product Code. 16879783
  • 145.00€ / 20µL
Description
This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis (Tiranti et al. 1995).
Specifications
Specifications
SSBP1
Polyclonal
Unconjugated
SSBP1
Mt SSB, MtSSB, PWP1 interacting protein 17, SSBP, SSBP1
Rabbit
Antigen Affinity Chromatography
RUO
381760, 54304, 6742
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28042, Q04837, Q9CYR0
Ssbp1
SSBP1 Fusion Protein Ag2860
20 μL
Primary
Rat, Human, Mouse