missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ssDNA Antibody (TNT-3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29849H
This item is not returnable.
View return policy
Description
ssDNA Monoclonal antibody specifically detects ssDNA in All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ssDNA | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TNT-3 | |
| Simple Western, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Necrotic marker, Single Stranded DNA | |
| Human Raji cell nuclei. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction