missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (CL6334), Novus Biologicals™
Mouse Monoclonal Antibody
369.00€ - 534.00€
Specifications
| Antigen | SSEA-1 |
|---|---|
| Clone | CL6334 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602990
|
Novus Biologicals
NBP2-76487-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366290
|
Novus Biologicals
NBP2-76487 |
100 μL |
534.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot.Tekniske data
| SSEA-1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| FUT4 | |
| IgG2a | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL6334 | |
| Monoclonal | |
| Purified | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 2526.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RKPSGAGWEKEWAEAPQEA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel