missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SSM 3 trifluoroacetate
GREENER_CHOICE

Product Code. 18711378
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
18711378 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 18711378

Brand: Tocris Bioscience™ 5253/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent furin inhibitor

SSM 3 trifluoroacetate is a potent furin inhibitor (EC50 = 54 nM). Blocks furin-dependent cell surface processing of anthrax protective antigen-83 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-N,N′′-[[(1R,3S,4S,6R)-4,6-Bis[(aminoiminomethyl)amino]-1,3-cyclohexanedyil]bis(oxy-4,1-phenylene)]bisguanidine tetratrifluoroacetate salt
CAS 922732-52-3
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C22H32N12O2.4CF3CO2H
Formula Weight 952.66
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.