Learn More
Biotin Antibody (BK-1/39), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50120MFV500
This item is not returnable.
View return policy
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for Immunohistochemistry
Specifications
| Biotin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry | |
| BK-1/39 | |
| Immunohistochemistry | |
| Vitamin B4 | |
| Biotin conjugated to keyhole limpet haemocyanin | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |