missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Mouse anti-Human, Clone: B-L5, Novus Biologicals™
Mouse Monoclonal Antibody
389.00€ - 736.00€
Specifications
| Antigen | ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 |
|---|---|
| Clone | B-L5 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, ELISA |
| Applications | Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18370865
|
Novus Biologicals
NBP3-18098-200UL |
200 μg |
389.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18395035
|
Novus Biologicals
NBP3-18098-500UL |
500 μg |
736.00€
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal antibody specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 | |
| 1 mg/ml | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Golgi Apparatus Markers | |
| PBS. Sterile-filtered through 0.22 um. | |
| 6480 | |
| IgM κ | |
| Affinity purified |
| B-L5 | |
| Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N | |
| Raji cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title