missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Stablcal Reference Suspension Set

Product Code. 10098114
Change view
Click to view available options
Includes:
100 mL bottle set: less than 0.1; 3; 6; 18; 30 NTU/FNU
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10098114 100 mL bottle set: less than 0.1; 3; 6; 18; 30 NTU/FNU 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10098114 Supplier Hach Lange™ Supplier No. 2897200
  • ONLINE EXCLUSIVE PRICE
    1140.83€
    943.03€ / 500mL
    Save 197.80€
    17% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Description Set of reference suspensions I, II, III, IV with Pharm. Eur. 5.0, 2100AN/AN IS with certificate
Includes 100 mL bottle set: less than 0.1; 3; 6; 18; 30 NTU/FNU
compliance-icons
Product Identifier
  • StablCal Formazin Turbidity Standard, < 0.1 NTU
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1A
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methenamine, Paraformaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.