Learn More
Eppendorf™ Stacking and Leveling
Product Code. 17171066
Change view
Click to view available options
For Use With (Application):
Stack and Level >180 kg
Stack and Level <100 kg
Stack and Level 100 to 180 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17171066 Stack and Level >180 kg Each
15713695 Stack and Level <100 kg Each
17161066 Stack and Level 100 to 180 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17171066 Supplier Eppendorf™ Supplier No. 2277000013
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stacking and leveling Incubators/Shakers
  • Stacking and leveling Incubators/Shakers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Incubators and Shakers
Product Type Stack and Level Equipment
For Use With (Application) Stack and Level >180 kg