Learn More
Rogo Sampaic™ Adson Stainless Steel Forceps
Product Code. 11762295
Change view
Click to view available options
Length (Metric):
120 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11762295 120 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11762295 Supplier Rogo Sampaic™ Supplier No. FML001
  • ONLINE EXCLUSIVE PRICE
    117.00€
    19.70€ / Pack of 10
    Save 97.30€
    83% Off
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
With small, interlocking teeth to help hold and grasp tissue. Rogo Sampaic™ Adson Stainless Steel Forceps are excellent for use on dressing materials such as cotton and gauze during surgical procedures.
  • 120mm in length
  • Supplied in packs of 10
Specifications
Product Type Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity 10/Pk.
Tip Style Fine, Round
Length (Metric) 120 mm