Learn More
Lenz Laborglasinstrumente™ Fork Clamps for Spherical Joints
Product Code. 10169550
Change view
Click to view available options
Size:
Spherical Joint: NS 13
Spherical Joint: NS 19
Spherical Joint: NS 29
Spherical Joint: NS 35
Spherical Joint: NS 40/41
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10169550 Spherical Joint: NS 19 Each
10463681 Spherical Joint: NS 13 Each
10342661 Spherical Joint: NS 35 Each
10444061 Spherical Joint: NS 29 Each
10382611 Spherical Joint: NS 40/41 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10169550 Supplier Lenz Laborglasinstrumente™ Supplier No. 01231909
  • ONLINE EXCLUSIVE PRICE
    19.13€
    12.91€ / Pack of 1
    Save 6.22€
    32% Off
In Stock
Add to basket
Edge
Add to basket
Ensures a secure connection between laboratory apparatus. Lenz Laborglasinstrumente™ Fork Clamps for Spherical Joints are manufactued from durable stainless steel.
  • With set screw
  • Available for a range of spherical joint sizes
Specifications
Quantity 1 Ea.
Type Joint Clamp
Material Stainless Steel
Includes Set Screw
For Use With (Equipment) For spherical joints
Size Spherical Joint: NS 19