Learn More
Rogo Sampaic™ Dumont™ No. 5 Stainless Steel Forceps
Description
- 110mm in length
- Supplied in packs of 10
Specifications
Specifications
| Product Type | Forceps |
| Material | Stainless Steel |
| For Use With (Application) | For dissection |
| Quantity | 10/Pk. |
| Tip Style | Sharp, Fine |
| Length (Metric) | 110 mm |