Learn More
Agencinox™ Stainless Steel Footstool
Product Code. 11709885
Change view
Click to view available options
Height (Metric):
180 mm
210 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11709885 210 mm Each
11744926 180 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11709885 Supplier Agencinox™ Supplier No. 759
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Use this footstool to safely retrieve items from high places. Agencinox™ Stainless Steel Footstools feature non-slip steps and are designed for use in laboratories and medical environments.
  • Anti-skidding steps
  • 18/10 Stainless steel construction
  • Available in a variety of sizes
Specifications
Height (Metric) 210 mm
Quantity 1 Each
Width (Metric) 340 mm
Material Stainless Steel
Product Type Foot Stool