Learn More
Nickel Electro™ Stainless Steel Forceps
Product Code. 15699310 Shop All Nickel-Electro Products
Change view
Click to view available options
Length (Metric):
300 mm
Unit Size:
1 pair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15699310 300 mm 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15699310 Supplier Nickel Electro™ Supplier No. 7117
  • ONLINE EXCLUSIVE PRICE
    12.70€
    8.51€ / 1 pairs
    Save 4.19€
    32% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Straight, blunt round ends. Nickel-Electro™ Stainless Steel Forceps contain serrated jaws.
  • Stainless Steel Forceps
  • Straight, serrated jaws
  • Blunt round ends
  • 300mm Length
Specifications
Product Type Laboratory forceps/tweezers
Length (English) 12 in.
Length (Metric) 300 mm
Material Stainless Steel
Dimensions (L x W) 300 mm
No. per Pack 1 Ea.
Tip Style Blunt