Specifications
| Type | Graphseal Ferrule |
| Diameter (Metric) Outside | 0.8 mm |
| For Use With (Application) | Capillary Column |
| For Use With (Equipment) | GC Capillary Columns |
| Material | Stainless Steel |
| Quantity | 10/Pk. |
| Diameter (Metric) Inside | 0.8 mm |