Learn More
Macherey-Nagel™ Stainless Steel Graphseal Ferrule
Product Code. 10718612 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inside:
0.4 mm
0.5 mm
0.8 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
10718612 0.8 mm Pack of 10
10481262 0.4 mm Pack of 10
10391722 0.5 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10718612 Supplier Macherey-Nagel™ Supplier No. 708319
Only null left
Add to basket
Add to basket
Specifications
Type Graphseal Ferrule
Diameter (Metric) Outside 0.8 mm
For Use With (Application) Capillary Column
For Use With (Equipment) GC Capillary Columns
Material Stainless Steel
Quantity 10/Pk.
Diameter (Metric) Inside 0.8 mm