Learn More
Carver™ Stainless Steel Heated Platens
Product Code. 11980017
Change view
Click to view available options
Dimensions (L x W):
6 x 6 in. Plate Size
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11980017 6 x 6 in. Plate Size Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Special packaging extra freight cost. Please contact us for quotation
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11980017 Supplier Carver™ Supplier No. 4286CE
  • ONLINE EXCLUSIVE PRICE
    23287.56€
    8361.99€ / Pack of 2
    Save 14925.57€
    64% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Special packaging extra freight cost. Please contact us for quotation
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Digitally controlled steel heated platens allow more precise control. Carver™ Stainless Steel Heated Platens are controlled automatically from a separate control panel.
  • Digital control for greater precision.
  • Individual temperature controllers automatically control platen temperature from a separate control panel.
  • Suitable for all Model C presses
Specifications
Voltage 230 V
For Use With (Equipment) Model C Presses
Dimensions (L x W) 6 x 6 in. Plate Size
Temperature Range (English) 650°F