Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Product Code. 10146383
Change view
Click to view available options
Needle Gauge:
22 G
22s G
23 G
23s G
25s G
26 G
26s G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Each
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Needle Point Style unitSize
10146383 22 G 5 or AS Pack of 6
11542525 22s G 2, 3, or 4 Pack of 6
10268047 23 G 2, 3, or 4 Pack of 6
10402205 22 G 2, 3, or 4 Pack of 6
12316309 23s G 5 or AS Pack of 6
15937335 26 G 2, 3, or 4 Each
11550042 26 G 2, 3, or 4 Pack of 6
10051173 26s G 2, 3, or 4 Pack of 6
12326459 25s G 2, 3, or 4 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10146383 Supplier Hamilton™ Supplier No. 773201
Estimated Shipment: 14-07-2026
Add to basket
Add to basket
Can be used with removable (RN)-style syringes that are 250μL and larger.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge 22 G
Diameter (Metric) Inner 0.41 mm
Diameter (Metric) Outer 0.72 mm
Product Type Removable Custom Needle
Quantity 6/Pk.
Needle Point Style 5 or AS
For Use With (Equipment) Removable Syringes (250μL and larger)