Learn More
Sampling Systems™ Stainless Steel Needle
Product Code. 17972012 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
145 mm
147 mm
Style:
Curved
Straight
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Style Unit Size
17972012 147 mm Straight Each
17932112 145 mm Curved Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17972012 Supplier Sampling Systems™ Supplier No. A638140
  • ONLINE EXCLUSIVE PRICE
    1.55€
    1.48€ / Pack of 1
    Save 0.07€
    4% Off
View all promotions
Estimated Shipment: 28-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This range of stainless steel needles makes light work of handling small and intricate materials.
  • Can be used to pick up small items or dissection
  • Available with either a straight end or curved end
  • Made from one piece and can be easily cleaned after use
  • Sharp point is ideal for use in laboratory and production work
Specifications
Product Type Dissection Needle
Color Silver
Material Stainless Steel (AISI 304)
Style Straight
Length (English) 5.8 in.
Length (Metric) 147 mm