Learn More
De Buyer™ Stainless Steel Pastry Bowls
Product Code. 11940635
Change view
Click to view available options
Volume (Metric):
1 L
3.5 L
4.5 L
5.5 L
7 L
8 L
11.4 L
16.8 L
Height (Metric):
90 mm
100 mm
112 mm
120 mm
125 mm
140 mm
145 mm
150 mm
200 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Height (Metric) unitSize
11940635 8 L 140 mm Each
11990625 1 L 100 mm Each
11980625 1 L 90 mm Each
11910605 3.5 L 112 mm Each
11910635 4.5 L 120 mm Each
11920635 5.5 L 125 mm Each
11950635 11.4 L 150 mm Each
11920645 7 L 145 mm Each
11940645 16.8 L 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11940635 Supplier De Buyer™ Supplier No. 3250.32
Specifications
Product Type Pastry Bowl
Material Stainless Steel
For Use With (Application) For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients
For Use With (Equipment) Fridge, deep-freezer, blast-chiller and oven
Height (Metric) 140 mm
Quantity Each
Shape Flat Bottom
Sterility Sterile
Volume (Metric) 8 L