Learn More
Kap-Uts Culture Tube Closures
Product Code. 15215637 Shop All Cole-Parmer Products
Change view
Click to view available options
Size (Metric):
13 mm
16 mm
Unit Size:
Pack of 144
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Unit Size
15215637 16 mm Pack of 144
15245376 13 mm Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15215637 Supplier Kap-Uts Supplier No. 0450001
  • ONLINE EXCLUSIVE PRICE
    825.00€
    772.06€ / Pack of 144
    Save 52.94€
    6% Off
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These Culture Tube Closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
Especificaciones
Product Type Culture Tube Closure
Autoclavable Autoclavable
Material Stainless Steel
Quantity 144 Pk.
Size (English) 0.6 in.
Size (Metric) 16 mm
Tube Material Stainless Steel
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.