Learn More
Endecotts™ Stainless Steel Receiver
Product Code. 12825691
Change view
Click to view available options
Diameter (Metric):
200 mm
230 mm
450 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12825691 450 mm Each
11537524 200 mm Each
10492065 230 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12825691 Supplier Endecotts™ Supplier No. 450S_STREC
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
  • Stainless steel construction
  • For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Product Type Bottom Shelter for Screening
Material Stainless Steel
Diameter (Metric) 450 mm
Packaging Quantity 1 Ea.