Learn More
Sampling Systems™ Stainless Steel Sample Splitter Riffle Boxes
Product Code. 15899531 Shop All Sampling Systems Products
Change view
Click to view available options
Width (Metric) Chute:
6 mm
10 mm
20 mm
30 mm
No. of Chutes:
12
16
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Chute No. of Chutes Unit Size
15899531 20 mm 12 Each
15809541 30 mm 12 Each
15889531 10 mm 16 Each
15879531 6 mm 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15899531 Supplier Sampling Systems™ Supplier No. 8601S20
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These easy-to-use Sample Splitters in stainless steel are for dividing and reducing bulk samples.
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifications
Product Type Riffle Box
Material Stainless Steel
No. of Chutes 12
For Use With (Application) Dividing and reducing bulk samples
Width (Metric) Chute 20 mm