Learn More
Rogo Sampaic™ Fixed Blade Scalpels
Product Code. 11708551
Change view
Click to view available options
Length (Metric) Blade:
40 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Unit Size
11708551 40 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708551 Supplier Rogo Sampaic™ Supplier No. FMB003
  • ONLINE EXCLUSIVE PRICE
    23.85€
    20.50€ / Pack of 10
    Save 3.35€
    14% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent for cutting and dissection needs in the laboratory. Rogo Sampaic™ Fixed Blade Scalpels are manufactured from durable, attractive stainless steel.
  • Handle length of 100mm
  • Fixed blade length of 40mm
  • Provided in packs of 10
Specifications
Blade Material Stainless Steel
Length (Metric) Blade 40 mm
Handle Material Metal