Learn More
Rogo Sampaic™ Stainless Steel Scissors
Product Code. 11796135
Change view
Click to view available options
Length (Metric):
110 mm
115 mm
120 mm
140 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
For Use With (Application):
Anatomical
Dissecting
Iris
Metzenbaum
Noyes micro
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style For Use With (Application) Unit Size
11796135 120 mm Round/Round Dissecting Pack of 10
11738531 140 mm Round/Round Anatomical Pack of 10
11768531 110 mm Sharp/Sharp Iris Pack of 10
11786135 140 mm Round/Round Metzenbaum Pack of 10
11748531 140 mm Sharp/Round Anatomical Pack of 10
11706145 115 mm Sharp/Sharp Noyes micro Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11796135 Supplier Rogo Sampaic™ Supplier No. FMN009
  • ONLINE EXCLUSIVE PRICE
    31.80€
    27.32€ / Pack of 10
    Save 4.48€
    14% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Excellent for a variety of cutting applications in the laboratory. Rogo Sampaic™ Stainless Steel Scissors have a beautiful polished matte finish.
  • Surgical grade stainless steel
  • Available in a variety of styles - Metzenbaum, dissecting, iris, anatomical
Specifications
Product Type Stainless Steel Scissors
Length (Metric) 120 mm
Material Stainless Steel
For Use With (Application) Dissecting
Tip Style Round/Round