Learn More
Aalborg™ Precision Valve for Stainless Steel Flowmeter
Product Code. 11778368
Change view
Click to view available options
Airflow Rate:
1000 mL/min.
200 mL/min.
21500 mL/min.
2500 mL/min.
400 mL/min.
Waterflow Rate:
12mL/min.
26mL/min.
650mL/min.
6mL/min.
80mL/min.
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Waterflow Rate unitSize
11778368 1000 mL/min. 26mL/min. Each
11758368 200 mL/min. 6mL/min. Each
11768368 400 mL/min. 12mL/min. Each
11788368 2500 mL/min. 80mL/min. Each
11708378 21500 mL/min. 650mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11778368 Supplier Aalborg™ Supplier No. VM3SV
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Aalborg™ Precision Valve for Stainless Steel Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • For high precision
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Shells are made of stainless steel, bushings are made of plated brass or 316 stainless steel.
Specifications
For Use With (Equipment) 150 mm Gas/Liquid Flowmeter
Type Standard Valve
Airflow Rate 1000 mL/min.
Waterflow Rate 26mL/min.