missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-ABC Monoclonal Antibody (W6/32), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen H020T03Y04
This item is not returnable.
View return policy
Description
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumor defense. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-ABC | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.09% sodium azide, pH 7.2 | |
| P01889, P04439, P10321 | |
| HLA-A, HLA-B, HLA-C | |
| Affinity chromatography | |
| RUO | |
| 3105, 3106, 3107 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG2a κ |
| Flow Cytometry | |
| W6/32 | |
| NovaFluor Yellow 660 | |
| HLA-C | |
| A29 protein, alpha chain of MHC class II, antigen presenting molecule, AS, Aw-33, Aw-74, B-4901, B-5001, Bw-47, Bw-50, class I MHC, HLA-B antigen, D6S204, epididymis secretory protein Li 83, H-2K, H2-K, H-2K(d), HEL-S-83, histocompatibility antigen, HLA A, HLA -B, HLA B-40011, HLA B44, HLA C, HLA class I antigen, HLA class I antigen HLA-B, HLA class I histocompatibility antigen C alpha chain, HLA class I histocompatibility antigen Cw-1 alpha chain, HLA class I histocompatibility antigen, A alpha chain, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-11 alpha chain-like, HLA class I histocompatibility antigen, A-30 alpha chain, HLA class I histocompatibility antigen, A-33 alpha chain, HLA class I histocompatibility antigen, A-74 alpha chain, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-47 alpha chain, HLA class I histocompatibility antigen, B-50 alpha chain, HLA class I histocompatibility antigen, B-82 alpha chain, HLA class I histocompatibility antigen, C alpha chain, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLA class I histocompatibility antigen, Cw-15 alpha chain, HLA class I histocompatibility antigen, Cw-18 alpha chain, HLA locus, HLA molecule, HLAA, HLA-A, HLA-A locus alpha 2 domain, HLA-A null, HLA-A11, HLA-A33, HLA-Aw33.1, HLAB, HLA-B, HLA-B 1502, HLA-B alpha chain, HLA-B alpha chain (B*2706), HLA-B alpha chain (B*5002), HLA-B alpha-chain, HLA-B*45ZJ, HLA-B15, HLA-B35, HLA-B35 antigen, HLA-B-3506, HLA-B39, HLA-B-3905, HLA-B49, HLA-B50, HLA-B55, HLA-B-5502, HLA-B-5602, HLA-B59, HLA-B61, HLA-Bw50 antigen, HLAC, HLA-C, HLA-C alpha chain, HLA-C alpha chain (HLA-Cw*15v), HLA-C alpha chain for Cw*18GB, HLA-C alpha-chain, HLA-C antigen, HLA-Cw, HLA-Cw*04GB, HLA-Cw*1702, HLA-Cw*18GB, HLA-Cw-1503, HLA-DQB1, HLA-DRB1, HLA-JY3, HLC-C, Human leukocyte antigen A, Human leukocyte antigen C, human leukocyte antigen-C alpha chain, hypothetical protein, lecocyte antigen B, leucocyte antigen A, Leukocyte Antigen, leukocyte antigen C, leukocyte antigen class I-A, leukocyte antigen class I-B, Leukoocyte Antigen, LOC100515902, LOW QUALITY PROTEIN: HLA class I histocompatibility antigen, A-11 alpha chain-like, lymphocyte antigen, major histocompatability complex B antigen, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, A, major histocompatibility complex, class I, B, major histocompatibility complex, class I, C, MCH class I antigen, MGC184092, MGC7052, MHC, MHC 1, MHC antigen, MHC clas I antigen, MHC Class 1, MHC class 1 antigen, MHC class I, MHC class I alpha chain, MHC class I anitgen, MHC class I anti, MHC Class I antigen, MHC class I antigen A*30, MHC class I antigen A*33, MHC class I antigen A*74, MHC class I antigen B*47, MHC class I antigen B*50, MHC class I antigen B*82, MHC class I antigen Cw*15, MHC class I antigen Cw*18, MHC class I antigen GN00104, MHC class I antigen heavy chain, MHC class I antigen heavy chain HLA-C, MHC class I antigen HLA-A heavy chain, MHC class I antigen HLA-A33, MHC class I antigen HLA-B alpha chain, MHC class I antigen HLA-B heavy chain, MHC class I antigen null protein, MHC class I antigen SHCHA, MHC class I antigene, MHC class I histocompatibility antigen, MHC class I human leukocyte antigen, MHC class I molecule, MHC class I protein, MHC class II antigen, MHC class l antigen, MHC classI antigen, MHC HLA-ABC, MHC HLA-B cell surface glycoprotein, MHC HLA-B transmembrane glycoprotein, MHC HLA-B27d, MHC HLA-B27-HS, MHC HLA-B39N, MHC I HLA, MHC1, PSORS1, SPDA1, surface antigen, truncated class I antigen, truncated MHC class I antigen | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction