Learn More
Socorex™ Stainless Steel Straight Drenchers
Product Code. 11888432
Change view
Click to view available options
Length (Metric):
25 mm
51 mm
102 mm
203 mm
Unit Size:
Pack of 2
Pack of 3
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11888432 25 mm Pack of 3
11838442 51 mm Pack of 3
11878442 102 mm Pack of 2
11873193 203 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11888432 Supplier Socorex™ Supplier No. 376.0925S
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use Socorex™ Stainless Steel Straight Drenchers for animal feeding and dosing. These precision-assembled stainless-steel feeding needles are ideal for the delivery of fluid and are reusable since they are autoclavable.
  • Stainless Steel
  • Various lengths to accommodate feeding needs
  • Autoclavable
  • Reusable
Specifications
Autoclavable Autoclavable
Product Type Drenching Cannula
Material Stainless Steel
Length (Metric) 25 mm