Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Millimeter Pore Sizes with ASTM E 11
Product Code. 10709512 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
34 product options available for selection
Product selection table with 34 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10709512 1.18 mm Each
10074751 1.4 mm Each
10288681 1.7 mm Each
10370762 100 mm Each
10065011 106 mm Each
10575062 11.2 mm Each
10781233 12.5 mm Each
10025051 125 mm Each
10515262 13.2 mm Each
10289161 16 mm Each
10515452 19 mm Each
10380812 1 mm Each
10218831 2.36 mm Each
10228831 2.8 mm Each
10525452 22.4 mm Each
10268541 25 mm Each
10447961 26.5 mm Each
10330812 2 mm Each
10288871 3.35 mm Each
10576022 31.5 mm Each
10310242 4.75 mm Each
10094901 37.5 mm Each
10136601 45 mm Each
10185971 4 mm Each
10075031 5.6 mm Each
10506222 50 mm Each
10298681 53 mm Each
10498721 6.3 mm Each
10408731 6.7 mm Each
10600272 63 mm Each
10268781 75 mm Each
10574872 8 mm Each
10196211 9.5 mm Each
10278781 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
34 options
Product Code. 10709512 Supplier RETSCH Supplier No. 60.132.001180
  • ONLINE EXCLUSIVE PRICE
    127.00€
    125.31€ / Pack of 1
    Save 1.69€
    1% Off
Estimated Shipment: 30-09-2026
Specifications
Quantity 1 Each
Pore Size 1.18 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Shaker AS 200
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel