missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50mmH, with Perforated Plates with Slotted Holes
Brand: RETSCH 60.180.002800
Additional Details : Weight : 1.00000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve | |
Certificate indicating Compliance |
For separation, fractioning, particle size determination | |
200 mm | |
2.8mm | |
ISO 5223 | |
50mm |