Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Product Code. 17998461
Change view
Click to view available options
For Use With (Equipment):
100 L Container
170 L Container
170 L Semicircular Container
220 L Container
220 L Semicircular Container
310 L Container
500 L Container
75 L Semicircular Container
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17998461 170 L Semicircular Container Each
17938721 75 L Semicircular Container Each
17978641 100 L Container Each
17918661 170 L Container Each
17988641 220 L Container Each
17958941 220 L Semicircular Container Each
17958441 310 L Container Each
17958741 500 L Container Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17998461 Supplier Gilac™ Supplier No. G194795
  • ONLINE EXCLUSIVE PRICE
    1811.57€
    1198.29€ / Pack of 1
    Save 613.28€
    33% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless Steel Trolley for Large Volume Container makes it easier to work at breast height. Equipped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
  • Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Makes it easier to work at breast height.
Specifications
Type Trolley
Holds Large Volume Containers
Load Bearing Capacity (Metric) 100 kg
Material Stainless Steel
Caster Diameter (Metric) 125 mm
For Use With (Equipment) 170 L Semicircular Container
Height (English) 35.4 in.
Height (Metric) 900 mm