Learn More
Vici Valco Instruments™ Stainless Steel Tubing Cutter
Product Code. 12772680
Encompass
Change view
Click to view available options
For Use With (Equipment):
1/16 in. and 1/8 in. SS Tubing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12772680 1/16 in. and 1/8 in. SS Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12772680 Supplier Vici Valco Instruments™ Supplier No. JR792
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Achieve smooth and uniform cutting of stainless steel tubing with this cutting wheel type tubing cutter. Vici Valco Instruments™ Stainless Steel Tubing Cutter is ideal for cutting 1/16 in. and 1/8 in. SS tubing.
  • For 1/16 in. and 1/8 in. SS tubing
  • Leaves smooth, uniform cuts
  • Minimal deburring and/or reaming required
  • Easy replacement of the cutting wheel
  • Creates cuts for tube-to-tube and tube-to-frit connections
  • Creates suitable cuts for zerod dead volume connections
Specifications
Type Tubing Cutter
For Use With (Equipment) 1/16 in. and 1/8 in. SS Tubing