Learn More
VACUUBRAND™ Stainless Steel Vacuum Tubing With Small Flanges KF
Click to view available options
Tubing Size:
60mm
80mm
100mm
130mm
Unit Size:
Each
Description
- Manufactured from stainless steel
- Flange and tube TIG welded
- Available in a range of sizes
Specifications
Specifications
| Material | Stainless Steel |
| Diameter (Metric) Outer | 10 mm |
| Packaging Quantity | 1 Each |
| Tubing Size | 60mm |
| Includes | Small flange |
| Product Type | Vacuum Tubing |